Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010011.3 | NP_034141.3 | 3 | 385 | ||
GenBank mRNA | AB018421.1 | - | 3 | 379 | ||
AK002528.1 | - | 3 | 386 | |||
AK098101.1 | - | 3 | 383 | |||
BC010747.1 | - | 3 | 370 | 4242518 | ||
BC051049.1 | - | 3 | 371 | 4234581 | ||
X69296.1 | - | 359 |
Gene Symbol: Cyp4a31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201640.3 | NP_964002.2 | 3 | 418 |