Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slirp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026958.3 | NP_081234.2 | 4 | 312 | ||
GenBank mRNA | AJ293625.1 | - | 3 | 684 | ||
AK007699.1 | - | 4 | 299 | |||
AK028746.1 | - | 290 | ||||
BC065396.1 | - | 3 | 684 | |||
BC100567.1 | - | 4 | 277 | |||
BC116739.1 | - | 4 | 299 | 40126058 | ||
BC116741.1 | - | 4 | 299 | 40126060 | ||
BU850988.1 | - | 4 | 315 | |||
CK372908.1 | - | 4 | 282 |