Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dido1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291432.1 | NP_001278361.1 | 4 | 1331 | ||
NM_001291433.1 | NP_001278362.1 | 4 | 1383 | |||
NM_011805.3 | NP_035935.2 | 4 | 1383 | |||
NM_175551.4 | NP_780760.2 | 4 | 1383 | |||
NM_177852.4 | NP_808520.2 | 4 | 1331 | |||
GenBank mRNA | AJ238332.1 | - | 4 | 1285 | ||
AK014422.1 | - | 4 | 1381 | |||
AK129117.1 | - | 4 | 1331 | |||
AY425951.1 | - | 4 | 1285 | |||
AY425952.1 | - | 4 | 1285 | |||
BC029110.1 | - | 5 | 1385 | |||
BC060249.1 | - | 4 | 1331 | |||
BC096662.1 | - | 4 | 1323 | |||
BC138712.1 | - | 3 | 1160 | 8861734 | ||
BC138713.1 | - | 3 | 1160 | 8861735 | ||
BC145401.1 | - | 3 | 1160 |