Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030002.2 | NP_001025173.1 | 2, 3 | 283 | ||
NM_001242442.1 | NP_001229371.1 | 2, 3 | 367 | |||
NM_001242443.1 | NP_001229372.1 | 2, 3 | 428 | |||
NM_005310.3 | NP_005301.2 | 2, 3 | 438 | |||
XM_017024536.1 | XP_016880025.1 | 2, 3 | 1376 | |||
XM_017024537.1 | XP_016880026.1 | 2, 3 | 405 | |||
XM_017024538.1 | XP_016880027.1 | 2, 3 | 424 | |||
GenBank mRNA | AB008789.1 | - | 1, 2 | 214 | ||
AB008790.1 | - | 1, 2 | 214 | |||
AK125544.1 | - | 2, 3 | 367 | |||
AK222849.1 | - | 2, 3 | 386 | |||
AK222870.1 | - | 2, 3 | 385 | |||
AK290115.1 | - | 2, 3 | 428 | |||
AK294620.1 | - | 2, 3 | 300 | |||
AK314368.1 | - | 2, 3 | 420 | |||
AU124875.1 | - | 2, 3 | 283 | |||
BC006535.2 | - | 2, 3 | 269 | 2824302 | ||
BT006686.1 | - | 1, 2 | 163 | |||
D43772.1 | - | 2, 3 | 382 | |||
D87513.1 | - | 1, 2 | 94 | |||
DC384178.1 | - | 2, 3 | 438 | |||
KU178103.1 | - | 1, 2 | 163 | |||
KU178104.1 | - | 1, 2 | 163 |