Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011758.2 | NP_001011758.2 | 1 | 444 |
Set Membership: |
G-protein coupled receptor |