Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276378.1 | NP_001263307.1 | 5 | 594 | ||
NM_001276379.1 | NP_001263308.1 | 4 | 469 | |||
NM_020347.3 | NP_065080.1 | 3 | 313 | |||
NR_075080.1 | 2 | 188 | ||||
XM_006713207.2 | XP_006713270.2 | 3 | 298 | |||
XM_011533838.2 | XP_011532140.1 | 3 | 237 | |||
XM_017006644.1 | XP_016862133.1 | 6 | 951 | |||
XM_017006645.1 | XP_016862134.1 | 5 | 826 | |||
GenBank mRNA | AJ297351.1 | - | 3 | 270 | ||
AK024323.1 | - | 3 | 264 | |||
AK093705.1 | - | 5 | 912 | |||
AK303416.1 | - | 6 | 365 | |||
BC025988.2 | - | 3 | 242 | 4838035 | ||
BC042483.1 | - | 3 | 273 | 4831113 | ||
BX640604.1 | - | 3 | 298 | |||
CN429018.1 | - | 2 | 97 | |||
DA408809.1 | - | 2 | 159 | |||
DC410047.1 | - | 4 | 469 | |||
HY159348.1 | - | 3 | 314 |