Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019866.1 | NP_063919.1 | 4 | 254 | ||
GenBank mRNA | BC141118.1 | - | 4 | 254 | 9055730 |