Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcbtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027764.2 | NP_082040.2 | 4, 5 | 842 | ||
GenBank mRNA | AK033501.1 | - | 4, 5 | 776 | ||
AK088299.1 | - | 4, 5 | 831 | |||
AK089923.1 | - | 5, 6 | 843 | |||
BC067005.1 | - | 5, 6 | 896 | 5721370 |