Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020369.2 | NP_065102.1 | 1 | 320 | ||
GenBank mRNA | AF281049.1 | - | 1 | 320 | ||
AK225867.1 | - | 1 | 274 | |||
AK300518.1 | - | 1 | 253 | |||
AK301765.1 | - | 1 | 224 | |||
AK314050.1 | - | 1 | 225 | |||
BC035606.1 | - | 1 | 195 | 5172588 |