Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pebp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028526.4 | NP_082802.1 | 2 | 274 | ||
NM_028560.4 | NP_082836.2 | 2 | 274 | |||
GenBank mRNA | AK006750.1 | - | 2 | 275 | ||
AK006964.1 | - | 2 | 265 | |||
BC048621.1 | - | 2 | 252 | 6742909 |