Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dppa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028615.1 | NP_082891.1 | 7 | 1405 | ||
GenBank mRNA | AF490346.1 | - | 7 | 1203 | ||
AK010743.1 | - | 7 | 1203 | |||
AK145645.1 | - | 7 | 1232 | |||
AK145760.1 | - | 7 | 1202 | |||
AK163375.1 | - | 7 | 1205 | |||
AK167211.1 | - | 7 | 1203 | |||
AY561246.1 | - | 7 | 1405 |