Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028878.3 | NP_083154.1 | 6 | 892 | ||
GenBank mRNA | AJ633679.1 | - | 6 | 901 | ||
AK014544.1 | - | 6 | 893 | |||
AK028784.1 | - | 6 | 893 | |||
AK128923.1 | - | 5 | 1235 | |||
AK162399.1 | - | 5 | 637 | |||
BC107411.1 | - | 6 | 893 | 40059042 | ||
BC107412.2 | - | 6 | 897 | 40059050 |