Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glt8d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029102.3 | NP_083378.2 | 10 | 1153 | ||
GenBank mRNA | AK003894.1 | - | 10 | 1154 | ||
BC082561.1 | - | 10 | 1082 | 30603848 |