Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020378.3 | NP_065111.1 | 3 | 1212 | ||
GenBank mRNA | AB038651.1 | - | 3 | 1196 | ||
AF086533.1 | - | 3 | 408 | |||
BC019079.2 | - | 3 | 1175 | 4560462 |