Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brsk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009929.3 | NP_001009929.1 | 10 | 1227 | ||
NM_001009930.3 | NP_001009930.1 | 10 | 1227 | |||
NM_001276763.1 | NP_001263692.1 | 10 | 1227 | |||
NM_029426.2 | NP_083702.1 | 10 | 1227 | |||
GenBank mRNA | AK158574.1 | - | 10 | 976 | ||
AK173268.1 | - | 9 | 802 | |||
AY533672.1 | - | 10 | 892 | |||
AY533673.1 | - | 10 | 892 | |||
AY533674.1 | - | 10 | 892 | |||
AY660739.1 | - | 10 | 1227 | |||
BC056498.1 | - | 7 | 503 | |||
JF707771.1 | - | 9 | 860 | |||
JF707772.1 | - | 10 | 892 |