Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crebbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025432.1 | NP_001020603.1 | 10 | 2190 | ||
GenBank mRNA | AK136057.1 | - | 5 | 618 | ||
BC072594.1 | - | 10 | 2176 |