Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC053393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025435.3 | NP_001020606.2 | 3 | 434 | ||
GenBank mRNA | AK140690.1 | - | 3 | 289 | ||
AK145729.1 | - | 3 | 310 | |||
AK166838.1 | - | 3 | 311 | |||
BC052651.1 | - | 3 | 425 | 30034019 | ||
BC053393.1 | - | 2618 |
Gene Symbol: Gm12169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163357.1 | NP_001156829.1 | 3 | 364 | ||
GenBank mRNA | BG079892.2 | - | 3 | 349 |