Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm12b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198957.2 | NP_945195.1 | 4 | 3358 | ||
GenBank mRNA | AK080943.1 | - | 4 | 3286 | ||
BB653094.1 | - | 1 | 432 | |||
BC030408.1 | - | 1 | 1695 | |||
BC080741.1 | - | 3 | 3272 | 30461213 | ||
BY620336.1 | - | 1 | 315 |