Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rusc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037709.2 | NP_001032798.1 | 12 | 5111 | ||
NM_199057.3 | NP_951012.2 | 12 | 5072 | |||
GenBank mRNA | AK053542.1 | - | 6 | 4416 | ||
AK122263.1 | - | 12 | 4991 | |||
AK132170.1 | - | 12 | 5001 | |||
AK166260.1 | - | 12 | 5006 | |||
BC024790.1 | - | 9 | 2384 | |||
BC141186.1 | - | 11 | 4836 | 9055798 |