Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eprs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029735.1 | NP_084011.1 | 15, 16 | 2061 | ||
GenBank mRNA | AK028687.1 | - | 8, 9 | 1082 | ||
AK141072.1 | - | 15, 16 | 2116 | |||
AK148463.1 | - | 15, 16 | 2022 | |||
AK160986.1 | - | 15, 16 | 2081 | |||
BC003226.1 | - | 3, 4 | 378 | |||
BC040802.1 | - | 15, 16 | 2059 | |||
BC058337.1 | - | 16, 17 | 2035 | |||
BC094679.1 | - | 14, 15 | 1855 | |||
BC141049.1 | - | 15, 16 | 1980 | 9055661 | ||
X54327.1 | - | 6, 7 | 745 |