Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eprs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029735.1 | NP_084011.1 | 4 | 411 | ||
GenBank mRNA | AK010734.1 | - | 4 | 375 | ||
AK088974.1 | - | 4 | 440 | |||
AK141072.1 | - | 4 | 463 | |||
AK147995.1 | - | 4 | 375 | |||
AK148463.1 | - | 4 | 372 | |||
AK160986.1 | - | 4 | 432 | |||
AK164073.1 | - | 3 | 2322 | |||
AK165637.1 | - | 3 | 204 | |||
AK201764.1 | - | 2 | 118 | |||
BC040802.1 | - | 4 | 409 | |||
BC058337.1 | - | 4 | 364 | |||
BC094679.1 | - | 3 | 205 | |||
BC141049.1 | - | 4 | 330 | 9055661 | ||
CB520692.1 | - | 3 | 223 | |||
CF537682.1 | - | 4 | 411 |