Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2010002M12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053217.2 | NP_444447.1 | 3 | 2071 | ||
GenBank mRNA | AK136525.1 | - | 2 | 1923 | ||
AK153247.1 | - | 3 | 2072 | |||
BC003493.1 | - | 2 | 888 | |||
BC138202.1 | - | 2 | 1902 | 8861221 |