Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127318.1 | NP_001120790.1 | 6 | 896 | ||
NM_145067.3 | NP_659504.2 | 6 | 896 | |||
GenBank mRNA | AK136496.1 | - | 6 | 920 | ||
BC099968.1 | - | 6 | 896 | 6432585 |