Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037712.1 | NP_001032801.1 | 5 | 970 | ||
GenBank mRNA | BC109174.1 | - | 5 | 970 | 40048550 | |
BC109175.1 | - | 5 | 969 |