Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252494.1 | NP_001239423.1 | 29 | 6877 | ||
NM_001252496.1 | NP_001239425.1 | 29 | 6598 | |||
NM_175258.4 | NP_780467.2 | 28 | 6862 | |||
GenBank mRNA | AK080138.1 | - | 1 | 935 | ||
AK164557.1 | - | 1 | 954 | |||
AK171771.1 | - | 2 | 2429 | |||
AK220459.1 | - | 17 | 4790 | |||
BC092525.1 | - | 9 | 3612 |