Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033218.1 | NP_150087.1 | 2 | 628 | ||
GenBank mRNA | AF374267.1 | - | 2 | 481 | ||
AK032847.1 | - | 2 | 652 | |||
AK053802.1 | - | 2 | 647 | |||
AK155857.1 | - | 3 | 576 | |||
AK172219.1 | - | 2 | 633 | |||
BC014730.1 | - | 2 | 618 | |||
BC023333.1 | - | 1 | 297 | |||
BC026556.1 | - | 2 | 620 | |||
BC038284.1 | - | 2 | 619 | |||
BC048071.1 | - | 2 | 633 | |||
BC069989.1 | - | 2 | 637 | |||
BQ965109.1 | - | 2 | 635 |