Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027748.3 | NP_082024.2 | 3 | 1000 | ||
GenBank mRNA | AJ292189.1 | - | 3 | 791 | ||
AK017128.1 | - | 1 | 1094 | |||
AK030878.1 | - | 1 | 69 | |||
AK033848.1 | - | 3 | 650 | |||
AK051249.1 | - | 1 | 371 | |||
AK154372.1 | - | 3 | 1009 | |||
BC039989.1 | - | 3 | 956 | |||
BC089030.1 | - | 3 | 839 | |||
BC094546.1 | - | 3 | 957 | |||
BC137617.1 | - | 3 | 941 | 8860636 | ||
BC137618.1 | - | 3 | 941 | 8860637 |