Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031811.2 | NP_001026981.2 | 9 | 1571 | ||
GenBank mRNA | AK048629.1 | - | 9 | 1460 | ||
BC029690.1 | - | 5 | 798 |