Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDR39U1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290292.1 | NP_001277221.1 | 5 | 825 | ||
NM_001290293.2 | NP_001277222.1 | 5 | 867 | |||
NM_001308075.1 | NP_001295004.1 | 5 | 787 | |||
NM_020195.2 | NP_064580.2 | 6 | 907 | |||
XM_011536975.1 | XP_011535277.1 | 6 | 951 | |||
XM_011536976.1 | XP_011535278.1 | 3 | 955 | |||
GenBank mRNA | AF226050.1 | - | 5 | 1181 | ||
AK123513.1 | - | 4 | 2014 | |||
BC000989.2 | - | 6 | 907 | 2900140 | ||
BC008566.1 | - | 5 | 1079 | |||
BX161384.1 | - | 4 | 818 |