Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il20rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033543.3 | NP_001028715.2 | 2 | 258 | ||
GenBank mRNA | BC107263.1 | - | 2 | 170 | ||
BC107264.2 | - | 2 | 170 | |||
BC148190.1 | - | 2 | 170 | |||
BY772000.1 | - | 2 | 260 | |||
CK331954.1 | - | 2 | 131 |