Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gls2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033264.3 | NP_001028436.2 | 10, 11 | 1086 | ||
NM_001285777.1 | NP_001272706.1 | 9, 10 | 1006 | |||
NM_001285779.1 | NP_001272708.1 | 10, 11 | 1115 | |||
GenBank mRNA | AK039618.1 | - | 9, 10 | 894 | ||
AK165896.1 | - | 10, 11 | 1032 | |||
AK168761.1 | - | 3, 4 | 218 | |||
AK220231.1 | - | 10, 11 | 1038 | |||
BC141115.1 | - | 10, 11 | 1086 | 9055727 | ||
EU770627.1 | - | 10, 11 | 1024 |