Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033126.2 | NP_001028298.1 | 1 | 220 | ||
NM_001042564.1 | NP_001036029.1 | 1 | 220 | |||
NM_001286753.1 | NP_001273682.1 | 1 | 220 | |||
GenBank mRNA | AA475779.1 | - | 1 | 147 | ||
AI326735.1 | - | 1 | 220 | |||
AK137770.1 | - | 1 | 197 | |||
AK153895.1 | - | 1 | 159 | |||
AK154005.1 | - | 1 | 204 | |||
BC139125.1 | - | 1 | 185 | 8862147 | ||
BC139126.1 | - | 1 | 185 | 8862148 | ||
BC171931.1 | - | 1 | 185 | 9054107 | ||
DT917063.1 | - | 1 | 146 | |||
DT924276.1 | - | 1 | 146 | |||
L24495.1 | - | 1 | 262 |