Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029983.2 | NP_001025154.1 | 11 | 1666 | ||
GenBank mRNA | AK088523.1 | - | 6 | 952 | ||
AK177212.1 | - | 2 | 175 | |||
AK193848.1 | - | 1 | 84 | |||
BC006645.1 | - | 9 | 1282 | |||
BC025500.1 | - | 3 | 318 | |||
BC138550.1 | - | 11 | 1622 | 8861570 | ||
BC138551.1 | - | 11 | 1622 | 8861571 |