Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029983.2 | NP_001025154.1 | 13 | 2819 | ||
GenBank mRNA | AK088523.1 | - | 8 | 2105 | ||
BC006645.1 | - | 11 | 2435 | |||
BC025500.1 | - | 5 | 1471 |