Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010576.3 | NP_034706.3 | 28 | 9685 | ||
GenBank mRNA | AK040125.1 | - | 1 | 1317 | ||
AK171822.1 | - | 1 | 919 |