Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc102a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033533.3 | NP_001028705.2 | 4 | 1151 | ||
GenBank mRNA | AK144530.1 | - | 4 | 1104 | ||
AK165668.1 | - | 4 | 1108 | |||
BC107374.2 | - | 3 | 608 | |||
BC107375.1 | - | 3 | 611 | |||
BC114992.1 | - | 3 | 613 | |||
BC139307.1 | - | 3 | 996 | 8862329 | ||
BC139308.1 | - | 3 | 996 | 8862330 |