Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033321.1 | NP_001028493.1 | 7 | 951 | ||
GenBank mRNA | AK030046.1 | - | 8 | 1040 | ||
AK155425.1 | - | 7 | 951 | |||
BC100402.1 | - | 7 | 935 | 30106220 | ||
BC132428.1 | - | 7 | 1041 | 40130705 | ||
BC132430.1 | - | 7 | 1041 | 40130707 |