Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319182.1 | NP_001306111.1 | 11 | 1741 | ||
NM_001319183.1 | NP_001306112.1 | 10 | 2027 | |||
NM_001319184.1 | NP_001306113.1 | 10 | 1881 | |||
NM_020216.3 | NP_064601.3 | 11 | 2030 | |||
XM_005245421.1 | XP_005245478.1 | 10 | 1548 | |||
XM_017002022.1 | XP_016857511.1 | 10 | 1715 | |||
GenBank mRNA | AJ242586.1 | - | 11 | 2031 | ||
AJ296161.1 | - | 11 | 2001 | |||
AK026644.1 | - | 8 | 1233 | |||
AK097372.1 | - | 12 | 1944 | |||
AL390139.1 | - | 10 | 1710 | |||
BC001064.2 | - | 11 | 2015 | 2823082 | ||
BC012166.1 | - | 11 | 2007 | 4636934 |