Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033339.3 | NP_001028511.1 | 4 | 1083 | ||
GenBank mRNA | AK089273.1 | - | 4 | 823 | ||
AK089533.1 | - | 4 | 823 | |||
AK137491.1 | - | 4 | 685 | |||
AK154458.1 | - | 4 | 1085 | |||
AK171493.1 | - | 4 | 823 | |||
BC059059.1 | - | 3 | 1153 | |||
BC112379.1 | - | 4 | 769 | 40059601 |