Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033339.3 | NP_001028511.1 | 2, 3 | 822 | ||
GenBank mRNA | AK089273.1 | - | 2, 3 | 562 | ||
AK089533.1 | - | 2, 3 | 562 | |||
AK137491.1 | - | 2, 3 | 530 | |||
AK154458.1 | - | 2, 3 | 824 | |||
AK171493.1 | - | 2, 3 | 562 | |||
BC059059.1 | - | 2, 3 | 796 | |||
BC112379.1 | - | 2, 3 | 508 | 40059601 |