Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033350.2 | NP_001028522.2 | 5 | 928 | ||
GenBank mRNA | AK041071.1 | - | 2 | 186 | ||
AK041242.1 | - | 5 | 889 | |||
AK156201.1 | - | 5 | 938 | |||
AY178734.1 | - | 5 | 948 | |||
BC026932.1 | - | 5 | 875 | 4008261 | ||
EU051378.1 | - | 4 | 529 |