Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033350.2 | NP_001028522.2 | 7 | 1163 | ||
GenBank mRNA | AK041071.1 | - | 4 | 421 | ||
AK041242.1 | - | 7 | 1124 | |||
AK156201.1 | - | 7 | 1173 | |||
AY178734.1 | - | 7 | 1183 | |||
BC026932.1 | - | 7 | 1110 | 4008261 | ||
EU051378.1 | - | 6 | 764 |