Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011988.2 | NP_036118.2 | 3 | 985 | ||
GenBank mRNA | AF072758.1 | - | 3 | 689 | ||
AJ577573.1 | - | 3 | 985 | |||
AK037042.1 | - | 3 | 745 | |||
AK076014.1 | - | 3 | 717 | |||
AK131156.1 | - | 2 | 1195 | |||
AK135190.1 | - | 3 | 962 | |||
BC060052.1 | - | 3 | 996 | |||
BC141134.1 | - | 3 | 950 | 9055746 |