Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53i11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025246.1 | NP_001020417.1 | 3 | 321 | ||
GenBank mRNA | AK142316.1 | - | 4 | 494 | ||
BC024710.1 | - | 2 | 290 | |||
BC048398.1 | - | 3 | 233 | 6397829 | ||
BC059041.1 | - | 3 | 193 | |||
BC098206.1 | - | 3 | 321 | 6812412 |