Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ott | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X96604.1 | - | 169 |
Gene Symbol: Gm10439 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037716.2 | NP_001032805.2 | 4 | 444 | ||
GenBank mRNA | BC109132.1 | - | 4 | 444 | 40038983 |