Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcrlb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029984.2 | NP_001025155.2 | 2, 3 | 57 | ||
GenBank mRNA | AY513660.1 | - | 2, 3 | 57 | ||
DQ119292.1 | - | 2, 3 | 57 |