Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015800.3 | NP_056615.1 | 17 | 3616 | ||
GenBank mRNA | AF168680.1 | - | 17 | 3068 | ||
AK034889.1 | - | 5 | 844 | |||
AK053197.1 | - | 15 | 2560 | |||
AK159603.1 | - | 4 | 495 | |||
BC100348.1 | - | 17 | 3401 | 30521148 |