Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030274.1 | NP_001025445.1 | 2 | 149 | ||
GenBank mRNA | AK148505.1 | - | 2 | 148 | ||
AK152845.1 | - | 2 | 144 | |||
AK186159.1 | - | 1 | 28 | |||
BC081434.1 | - | 2 | 93 | 6412214 | ||
BU534760.1 | - | 2 | 152 |