Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033135.3 | NP_001028307.2 | 2 | 571 | ||
GenBank mRNA | AK155360.1 | - | 2 | 630 | ||
AY155439.1 | - | 2 | 163 | |||
BC115968.1 | - | 3 | 151 | 40107454 | ||
BF018252.1 | - | 2 | 320 | |||
BY743657.1 | - | 2 | 571 |