Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kansl1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122738.1 | NP_001116210.1 | 2 | 690 | ||
NM_177645.4 | NP_808313.3 | 2 | 690 | |||
GenBank mRNA | AK081413.1 | - | 3 | 1040 | ||
AK138707.1 | - | 1 | 521 | |||
AK145440.1 | - | 2 | 661 | |||
AK170981.1 | - | 2 | 719 | |||
AK172645.1 | - | 2 | 643 | |||
AK220534.1 | - | 2 | 641 | |||
BC141036.1 | - | 3 | 600 | 9055648 |